missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ AgPath-ID™ One-Step RT-PCR Reagents with manual

Product Code. 30057764 Shop All Applied Biosystems Products
Change view
Click to view available options
No. of Reactions:
100 Reactions
1000 Reactions
500 Reactions
Unit Size:
100 reactions
1000 reactions
500 reactions
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions unitSize
30057764 500 Reactions 500 reactions
30057784 1000 Reactions 1000 reactions
17153279 100 Reactions 100 reactions
30057797 100 Reactions 100 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 30057764 Supplier Applied Biosystems™ Supplier No. 4387424M

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

AgPath-ID™ One-Step RT-PCR Reagents are designed for sensitive, robust amplification of RNA targets using a rapid single-tube TaqMan™ real-time reverse transcription PCR (RT-PCR) strategy.

  • Recommended for RNA pathogen amplification
  • Consistent amplification of RNA targets with high specificity and sensitivity
  • Optimized to work with your target-specific primers and probes
  • Contains ROX for quantitative fluorescent signal normalization

AgPath-ID™ One-Step RT-PCR Reagents are configured for fast and simple reaction setup. The reactions are assembled in a single tube, minimizing sample handling errors and expediting set-up time. Once assembled, results are available in approximately one hour. The 25X RT-PCR Enzyme Mix included in the kit contains highly efficient ArrayScript™ Reverse Transcriptase, a mutant MMLV RT that produces high cDNA yields, and AmpliTaq Gold™ polymerase, the preferred hot-start DNA polymerase for specific target amplification. The 2X RT-PCR Buffer has been optimized for efficient, robust reverse transcription and PCR includes the passive reference dye, ROX™ Dye, for quantitative fluorescent signal normalization.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 25X
Content And Storage Sufficient for 500 reactions of 25 μL, contains:

• 7 mL 2 x RT-PCR Buffer

• 550 μL 25 x RT-PCR Enzyme Mix

• 25 mL Nuclease-free Water.
Description Works on all AB platforms, but is optimized for the 7500.
Detection Method Primer-probe
Format Tube
GC-Rich PCR Performance High
PCR Method 1-step RT-qPCR
Polymerase AmpliTaq Gold DNA Polymerase
Reaction Speed Standard
For Use With (Equipment) 7500 System
Passive Reference Dye ROX (Pre-mixed)
Product Line AgPath-ID, Ambion
Product Type One-Step RT-PCR Reagent
Quantity 500 Reactions
Reverse Transcriptase ArrayScript™
Sufficient For 500 Reactions
For Use With (Application) Animal Health, Pathogen Detection
Fidelity (vs. Taq) 25 X
No. of Reactions 500 Reactions
Volume 7 mL
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.