missing translation for 'onlineSavingsMsg'
Learn More

Ion Torrent™ AgriSeq™ ULTRA Dual Barcode Adapters Combo Kit

Product Code. 30274962
Change view
Click to view available options
Quantity:
9600 Reactions
Unit Size:
9600 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30274962 9600 Reactions 9600 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30274962 Supplier Ion Torrent™ Supplier No. A40001408

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The AgriSeq ULTRA Dual Barcodes Adapters Combo Kit provides 384 unique START barcode adapters designed for optimal performance with 8 unique END barcode adapters, resulting in 3072 combinations for Ion Torrent semiconductor sequencers.

The AgriSeq ULTRA Dual Barcodes Adapters Combo Kit provides 384 unique START barcode adapters designed for optimal performance with 8 unique END barcode adapters, resulting in 3072 combinations for Ion Torrent semiconductor sequencers. When used with the AgriSeq ULTRA Library Kit, users can pool up to 3072 amplicon libraries and then conduct multiplexed sequencing analysis to help increase lab efficiency and reduce sequencing costs per sample.

Key product features:

  • Robust barcodes attach efficiently and uniformly to facilitate balanced representation of libraries
  • Unbiased performance enables identical genotypes independent of barcode used
  • Robust error correction built in by design for added confidence in sample identification

Increase sample multiplexing without impacting data quality

Multiplexing with barcoded libraries enables more cost effective runs by permitting up to 3072 unique samples per run, helping to reduce the cost and handling requirements of sequencing.

Each barcode was individually evaluated to verify no bias of sequence context representation. The barcodes were also screened for equivalent efficiency to verify balanced read depth between libraries. Regardless of the specific barcode used, AgriSeq targeted genotyping-by-sequencing (GBS) generates highly uniform libraries and concordant genotype calls.

TRUSTED_SUSTAINABILITY

Specifications

Format Kit
Isolation Technology Enzymatic PCR Clean-up Technology
Sequencing Type Next Generation Sequencing
For Use With (Equipment) Ion Torrent
For Use With (Application) Sequencing
Includes Combo Kit: 4 START Plates, 8 END Tubes
No. of Reactions 9600
Product Type Oligos - Barcodes
Quantity 9600 Reactions
Shipping Condition Cool temp

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.