missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ AHCYL1/CEN1q (Texas Red/FITC) FISH Probe

Product Code. 16093245
Change view
Click to view available options
Quantity:
100 μL
200 μL
Unit Size:
100µL
200µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16093245 100 μL 100µL
16003255 200 μL 200µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16093245 Supplier Abnova™ Supplier No. FG0157.100uL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe

Labeled FISH probes for identification of gene amplification using Fluoresecent In Situ Hybridization Technique.

FISH is a technique used to identify and localize the presence or absence of specific DNA sequences on cells and tissues. Abnova has developed a range of FISH probes for the detection of gene amplification, loss, and translocation. Each FISH probe has a pair of locus-specific, fluorophore-labeled probes originated from a bacterial chromosome (BAC) library.

Advantages

  • Dual colored probes for fast, sensitive, and specific detection
  • Work on metaphase spread, paraffin embedded, and frozen tissue
  • Identify gene amplification, loss, and translocation
  • High signal to noise ratio
  • Low cross reactivity

Specifications

Content And Storage Store at 4°C in the dark.
Description Labeled FISH probes for identification of gene amplification using Fluoresecent In Situ Hybridization Technique.
Format Tube
Label or Dye FITC, Texas Red
For Use With (Application) Fluorescent In Situ Hybridization (Cell)
Includes DAPI Counterstain (1500 ng/mL ) 250 uL
Location 1p13/1q21.3
Probe Approx. 150kb/600kb
Product Type AHCYL1/CEN1q FISH Probe
Quantity 100 μL
Source Genomic DNA
Species Human
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.