missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ AICAR
GREENER_CHOICE

Product Code. 18075772
missing translation for 'orderingAttributeHoverText'
Quantity:
50 mg
missing translation for 'unitSize'
50mg
This item is not returnable. View return policy

Product Code. 18075772

missing translation for 'mfr': Tocris Bioscience™ 2840/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

AMPK activator

Cell-permeable, allosteric activator of AMP-activated protein kinase (AMPK). Augments proliferation, differentiation and mineralization of osteoblastic MC3T3-EI cells and attenuates psychosine-induced expression of proinflammatory cytokines and iNOS in astrocytes. Promotes osteogenic differentiation of hAMSCs and BM-MSCs in vitro.
TRUSTED_SUSTAINABILITY

Specifications

CAS 2627-69-2
Quantity 50 mg
Molecular Formula C9H14N4O5
Formula Weight 258.23
Purity >99%
Molecular Weight (g/mol) 258.23
Chemical Name or Material AICAR
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.