missing translation for 'onlineSavingsMsg'
Learn More

AIM-100, MedChemExpress

Product Code. 16482527
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16482527 50 mg 50mg
16446716 100 mg 100mg
16426428 10 mg 10mg
16419017 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16482527 Supplier Medchem Express Supplier No. HY15290
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress AIM-100 is a potent and selective Ack1 inhibitor with an IC50 of 21.58 nM. AIM-100 also inhibits Tyr267 phosphorylation. AIM-100 does not inhibits other kinases including PI3-kinase and AKT subfamily members. AIM-100 has an anticancer effect.

AIM-100 is a potent and selective Ack1 inhibitor with an IC50 of 21.58 nM. AIM-100 also inhibits Tyr267 phosphorylation. AIM-100 does not inhibits other kinases including PI3-kinase and AKT subfamily members. AIM-100 has an anticancer effect.

Specifications

Chemical Name or Material AIM-100
CAS 873305-35-2
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C23H21N3O2
Quantity 50 mg
Solubility Information DMSO : 50 mg/mL (134.61 mM; Need ultrasonic)
SMILES C1(C2=C(C3=CC=CC=C3)OC4=C2C(NC[C@@H]5CCCO5)=NC=N4)=CC=CC=C1
Molecular Weight (g/mol) 371.43
Formula Weight 371.43
Percent Purity 99.92%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.