Learn More
BINDER™ Measurement of Air Exchange Rate Instrument
Product Code. 15631686 Shop All BINDER Products
Change view
Click to view available options
Product Type:
Air Exchange Measurement
Unit Size:
Each
Product Code. Product Type Unit Size
15631686 Air Exchange Measurement Each
1 options
Product Code. 15631686 Supplier BINDER™ Supplier No. 80120195
  • ONLINE EXCLUSIVE PRICE
    1325.00 €
    1261.00 € / Pack of 1
    Save 64.00 €
    4% Off
Estimated Shipment: 15-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
BINDER™ Measurement of Air Exchange Rate Instrument is designed according to ASTM D5374.
  • Accessory for Drying and heating chamber
  • Measures exchange rate in ambient temperature
  • Measurement in factory
  • With definition and protocol according to ambient temperature
  • For use with all FB and M models
Specifications
Product Type Air Exchange Measurement