Learn More
Abeba™ Air-Cushion Sole 2370 Shoes
Product Code. 15618127
Change view
Click to view available options
Size:
35
36
37
38
39
40
41
42
43
44
45
46
47
Unit Size:
1 pair
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
15618127 35 1 pair
15628127 36 1 pair
15638127 37 1 pair
15648127 38 1 pair
15658127 39 1 pair
15668127 40 1 pair
15678127 41 1 pair
15688127 42 1 pair
15698127 43 1 pair
15608137 44 1 pair
15618137 45 1 pair
15628137 46 1 pair
15638137 47 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Product Code. 15618127 Supplier Abeba™ Supplier No. 2370/35
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Antistatic with air cushion PU-sole and high slip resistance. Sanitized™ treated.
  • CE EN ISO 20347:2012 O1 SRA
  • Microfiber
  • White
  • Slip resistance PU outsole sole
  • Integrated footbed
  • Sanitized treated
  • Antistatic
  • Instep area with elastic band
  • Decontaminable
Specifications
Certifications/Compliance CE EN ISO 20347:2012 O1 SRA
Color White
Material Microfiber
Size 35
Sole Features Slip-resistant
Sole Material Polyurethane
Style 2370