missing translation for 'onlineSavingsMsg'
Learn More

Tork Air Freshener Spray Refill for A1 System

Product Code. 30243775 Shop All Tork Products
Change view
Click to view available options
Scent:
Citrus
Variety Pack
Unit Size:
Pack of 12
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Scent Unit Size
30243775 Citrus Pack of 12
30243874 Variety Pack Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30243775 Supplier Tork Supplier No. 236050
Safety Starts Here: Save Up to 60%
Use promo code 28008
View all promotions
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Control odors effectively and efficiently in your restroom with the Tork Air Freshener Spray.

  • Flexible setting to fit a variety of restrooms
  • Fresh citrus scent for a clean impression

Available scents:

  • Citrus
  • Variety pack, including Citrus, Fruit, and Floral
TRUSTED_SUSTAINABILITY

Specifications

Product Type Air Freshener Refill
Alternate Part Number 7322540030082
Color-coded No
Form Spray
Scent Citrus
Color Clear
Packaging Metal Can
Quantity 12/Pk.
Volume (Metric) 75 mL
compliance-icons compliance-icons
Product Identifier
  • Tork Citrus Air Freshener Spray / Tork Aérosol désodorisant Citron
Signal Word
  • Danger
Hazard Category
  • Flammable and non-flammable aerosols category 1
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H222-Extremely flammable aerosol.
  • H319-Causes serious eye irritation.
  • EUH208-Contains . May produce an allergic reaction.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P211-Do not spray on an open flame or other ignition source.
  • P251-Do not pierce or burn, even after use.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P410+P412-Protect from sunlight Do no expose to temperatures exceeding 50°C/122°F.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.