missing translation for 'onlineSavingsMsg'
Learn More

Kimberly-Clark Professional™ Rhapsodie Fragrance Aircare Refill

Product Code. 12641366 Shop All Kimberly-Clark Professional Products
Change view
Click to view available options
Scent:
Citrus
Unit Size:
Pack of 6
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Scent unitSize
12641366 Citrus Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12641366 Supplier Kimberly-Clark Professional™ Supplier No. 6136

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Eliminate unwanted smells and odors. Kimberly-Clark Professional™ RHAPSODIE Fragrance Aircare Refills are enriched with smooth, citrus notes promoting a pleasant washroom environment.

  • Supplied in packs of 6 x 310mL cassettes
  • Smooth, citrus notes
  • Contoured/rounded design prevents dust from being trapped in crevices
  • Compatible with the discontinued RIPPLE™ or AQUA™ Aircare Dispensers
TRUSTED_SUSTAINABILITY

Specifications

Scent Citrus
Appearance Clear
Format Refill
For Use With (Application) Aqua Aircare Dispenser
Quantity 6/Pk.
Volume (Metric) 310 mL
compliance-icons compliance-icons
Product Identifier
  • RHAPSODIE Fragrance Refill
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P370+P378-In case of fire: Use to extinguish.
  • P403+P235-Store in a well-ventilated place. Keep cool.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : propan-2-ol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.