Learn More
Asecos™ Air Purifier PURIFIAIR.488
Product Code. 17200197 Shop All asecos Products
Change view
Click to view available options
Product Type:
Air Purifier
Unit Size:
Each
Product Code. Product Type Unit Size
17200197 Air Purifier Each
1 options
Product Code. 17200197 Supplier Asecos™ Supplier No. 39311
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Air Purifier PURIFIAIR.488 reduces indoor pollutant levels with a multi-component filter system.
  • Asecos PURIFIAIR.488 can be used to reduce pollutant levels in indoor atmospheres
    • Removes mould spores, pollen, and allergens
    • Eliminates germs and bacteria and neutralises them
    • Continuously filters out fine dust particles
    • Removes unpleasant odours and harmful gases from indoor atmospheres
    • Built-in PM2.5 sensor and colour display allow users to quickly identify and easily read off the particle concentration in the indoor atmosphere in four levels from green (low) to red (high)
  • 1 device handles a volume flow rate of up to 488 m3/h and is designed for approx. 80 m3 room volume (corresponding to a surface area of 32 m2; at a room height of 2.5 m)
    • A larger number of devices should be used in larger rooms
    • Fresh air must be supplied to the rooms via natural or technical ventilation
    • Air purifiers should not be considered a substitute for a fresh air supply
    • Efficient results over time only if the filter is replaced at regularly according to the electronic display
    • Large filter units ensure a long service life
    • Low energy requirements result in low usage costs
    • Integrated humidification for a healthy and comfortable indoor climate
Specifications
Filter Type Prefilter, Cold Catalysis Filter, Activated Carbon Filter, High-performance Particle Filter
Airflow Rate 488 3/h
Weight (Metric) 11.3 Kg
Height (Metric) 669 mm
Depth (Metric) 400 mm
Width (Metric) 400 mm
Product Type Air Purifier
Voltage 220/240 V
Frequency 50/60 Hz
Certifications/Compliance CE Conformity