Learn More
Hamilton AirShield Tubing
Product Code. 31257789
Change view
Click to view available options
Length (English):
9.8 ft.
Product Code. Length (English)
31257789 9.8 ft.
1 options
Product Code. 31257789 Supplier Hamilton Supplier No. 9493901
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
AirShield tubing designed for use with compatible systems. Supports efficient airflow and reliable operation in laboratory setups.
  • Designed for demanding laboratory environments requiring enhanced protection
  • Snorkel connects directly to the AirShield to supply clean external air
  • Delivers clean air from outside the immediate workspace
  • Creates a protective airflow around critical instrument components
  • Helps reduce exposure to contaminants and improve system reliability
Specifications
Product Type AirShield Tubing Kit
Length (English) 9.8 ft.