missing translation for 'onlineSavingsMsg'
Learn More

AKBA, MedChemExpress

Product Code. 16469168
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16469168 10 mg 10mg
16461578 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16469168 Supplier Medchem Express Supplier No. HYN0892
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress AKBA (Acetyl-11-keto-β-boswellic acid) is an active triterpenoid compound from the extract of Boswellia serrate and a novel Nrf2 activator.

AKBA (Acetyl-11-keto-β-boswellic acid) is an active triterpenoid compound from the extract of Boswellia serrate and a novel Nrf2 activator.

Specifications

Chemical Name or Material AKBA
CAS 67416-61-9
Color White
Purity Grade Notes Research
Molecular Formula C32H48O5
Quantity 10 mg
Synonym Acetyl-11-keto-β-boswellic acid
Solubility Information DMSO : ≥ 5.2 mg/mL (10.14 mM)
SMILES C[C@@]([C@@](C1=C2)(CC[C@@](C)(CC[C@H]3C)[C@@]1([H])[C@H]3C)C)(CC[C@@]4([H])[C@@]5(C)C(O)=O)[C@@](C2=O)([H])[C@]4(CC[C@H]5OC(C)=O)C
Molecular Weight (g/mol) 512.72
Formula Weight 512.72
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.