Learn More
Eurofins DiscoverX Akt1/PKBα Protein, Unactive
Description
Akt1 is an oncogenic AGC kinase that plays a critical role in regulating many processes including cell survival, metabolism, proliferation, growth and angiogenesis. AKT activation relies on the PI3K pathway, and is recognized as a critical node in the pathway. The E17 hotspot is the most characterized of AKT1 mutations, and has been shown to result in activation of the protein. Mutations in AKT1 have also been shown to confer resistance to allosteric kinase inhibitors in vitro.
GenBank M63167. The recombinant protein contains the amino acid substitution S478G with respect to this accession number. This conflict is reported inEMBL BE 206796. The residue coordinates in the native sequence are given.
Specifications
Specifications
| Formulation | mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 150 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 50% glycerol, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. |
| Molecular Weight (g/mol) | 59 kDa |
| Quantity | 50 μg |
| Species | Human |
| Recombinant | Recombinant |
| Protein Tag | N-terminal 6His |
| Expression System | Recombinant enzyme expressed in Sf21 insect cells |
| Protein Form | Full-length |
| Purity or Quality Grade | ≥65% |
| Protein | Akt1/PKBa, unactive |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.