missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human AKT3 (PKB gamma), His Tag Recombinant Protein
Human AKT3 (PKB gamma), His Tag Recombinant Protein
Brand: Invitrogen PV3185
Additional Details : Weight : 3.36000kg
Description
AKT3 (PKB gamma) has been shown to be stimulated by platelet-derived growth factor (PDGF), insulin, and insulin-like growth factor 1 (IGF1). It is involved in a wide variety of biological processes including cell proliferation, differentiation, apoptosis, tumorigenesis, glycogen synthesis and glucose uptake.Specifications
NP_005456 | |
10000 | |
10μg | |
AKT Kinase Family | |
Recombinant | |
Storage Buffer: 20mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.01% Triton X-100, 0.1mM Na3VO4, 2mM DTT, 20% glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Ser⁄Thr Kinases: ACG Group | |
LanthaScreen™ Antibody, LanthaScreen™ Substrate, Z'-LYTE™, Omnia™ Bulk Substrate | |
v-akt murine thymoma viral oncogene homolog 3 | |
Kinases & Inhibitors | |
Liquid | |
Signal Transduction |
Kinase Assay | |
60.4 kDa | |
AKT3 (PKB gamma) | |
Human | |
His Tag | |
Biochemical terbium phospho-specific activity, Biochemical phospho-specific activity (no antibody) | |
Dry Ice | |
AKT3 | |
Baculovirus | |
Recombinant, Full Length | |
Serine/Threonine Kinases |