Learn More
Tocris Bioscience™ Akti-1/2
GREENER_CHOICE
Potent and selective dual Akt1 and 2 inhibitor
Brand:  Tocris Bioscience™ 5773/50
Encompass
Product Code. 15747106
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Akti-1/2 is a potent and selective dual Akt1 and 2 inhibitor (IC50 values are 50 and 210 nM, respectively). Selective for Akt1 and 2 over a panel of other tyrosine and serine/threonine kinases. Sensitizes LnCaP cells to TRAIL (TNF-related apoptosis-inducing ligand) induced apoptosis. Also enhances CAR and TCR retroviral transduction of human T cells. Active in vivo.
Specifications
Specifications
1,3-Dihydro-1-[1-[[4-(6-phenyl-1 H-imidazo[4,5-g]quinoxalin-7-yl)phenyl]methyl]-4-piperidinyl]-2 H-benzimidazol-2-one
612847-09-3
Akt Inhibitor VIII
50 mg
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.
Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato