missing translation for 'onlineSavingsMsg'
Learn More

Al-23 Boat, L1: 58mm, L2: 46mm, B1: 23mm, B2: 19mm, H: 13mm, W: 3mm, Vol.: 4.3, Thermo Scientific™

Product Code. 15493555
Change view
Click to view available options
Quantity:
1 Pc.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15493555 1 Pc. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15493555 Supplier Thermo Scientific Alfa Aesar Supplier No. 033173.KF
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Al-23
Quantity 1 Pc.
Form Boat
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.