Learn More
Al-23 Combustion Tray;L1 (mm), 100;L2 (mm), 98;B1 (mm), 50;B2 (mm), 48;H (mm), 30;W (mm), 4;Vol, 100.0
Product Code. 15403575
Change view
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Product Code. Quantity unitSize
15403575 1 Ea. Each
1 options
Product Code. 15403575 Supplier Thermo Scientific Alfa Aesar Supplier No. 033186.KF
 Restricted Item
In Stock
Add to basket
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Al-23 Combustion Tray
Quantity 1 Ea.
What is the maximum working temperature of Al-23 Tubes?
The maximum working temperature for the Al-23 Tube is 1950 degrees C.