missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals beta-Alanine, 98.5%, Thermo Scientific Chemicals

Product Code. 10167713 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
10 kg
Packaging:
Plastic Bottle
Plastic Drum
Unit Size:
10kg
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10167713 10 kg Plastic Drum 10kg
10245880 500 g Plastic Bottle 500g
10488890 2.5 kg Plastic Bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10167713 Supplier Thermo Scientific Chemicals Supplier No. 163790100
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 107-95-9 | C3H7NO2 | 89.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-95-9
Molecular Formula C3H7NO2
Molecular Weight (g/mol) 89.09
MDL Number MFCD00008200
InChI Key UCMIRNVEIXFBKS-UHFFFAOYSA-N
Synonym beta-alanine, 3-aminopropionic acid, abufene, beta alanine, h-beta-ala-oh, beta-aminopropionic acid, 2-carboxyethylamine, .beta.-alanine, alanine, beta, propanoic acid, 3-amino
PubChem CID 239
ChEBI CHEBI:16958
IUPAC Name 3-aminopropanoic acid
SMILES NCCC(O)=O

Specifications

Melting Point 197.0°C to 202.0°C
Ammonium 1000ppm max.
CAS Min % 98.5
CAS Max % 100.0
pH 6.5 to 7.5 (10% w/v aq. soln.)
Color White
Chlorine (Cl) 400ppm max.
Heavy Metals (as Pb) 10ppm max.
Infrared Spectrum Authentic
Iron (Fe) 30ppm max.
Assay Percent Range 99%
Loss on Drying 0.5% max. (105°C, 3 hrs)
Other Amino Acids (TLC) Passes Test (not detectable on 10μgspotting)
Packaging Plastic Drum
Linear Formula H2NCH2CH2CO2H
Fieser 01,16; 05,6; 06,10; 17,3
Sulfate 480ppm max.
Sulfated Ash 0.2% max.
Quantity 10 kg
Formula Weight 89.09
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material β-Alanine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.