missing translation for 'onlineSavingsMsg'
Learn More

Alantolactone, Tocris Bioscience™
GREENER_CHOICE

Product Code. 18014452
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18014452 10 mg 10mg
18004782 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18014452 Supplier Tocris Bioscience Supplier No. 4490/10

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Antiproliferative; induces activin/Smad3 signaling

Sesquiterpene lactone. Induces activin/SMAD3 signaling and disrupts Cripto-1/activin receptor type II A interaction. Displays antiproliferative activity in human colon adenocarcinoma HCT-8 cells.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 546-43-0
Molecular Formula C15H20O2
Molecular Weight (g/mol) 232.32
MDL Number MFCD00274568
InChI Key PXOYOCNNSUAQNS-AGNJHWRGSA-N
Synonym alantolactone, helenine, eupatal, alant camphor, inula camphor, helenin, elecampane camphor, unii-m7gsn5q1m6, m7gsn5q1m6, 3ar,5s,8ar,9ar-5,8a-dimethyl-3-methylidene-3a,5,6,7,8,8a,9,9a-octahydronaphtho 2,3-b furan-2 3h-one
PubChem CID 72724
ChEBI CHEBI:2540
IUPAC Name (3aR,5S,8aR,9aR)-5,8a-dimethyl-3-methylidene-2H,3H,3aH,5H,6H,7H,8H,8aH,9H,9aH-naphtho[2,3-b]furan-2-one
SMILES C[C@H]1CCC[C@]2(C)C[C@H]3OC(=O)C(=C)[C@H]3C=C12

Specifications

Quantity 10 mg
Formula Weight 232.32
Percent Purity >98%
Chemical Name or Material Alantolactone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.