missing translation for 'onlineSavingsMsg'
Learn More

Alarelin Acetate, MedChemExpress

Product Code. 16417328
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16417328 10 mg 10mg
16477286 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16417328 Supplier Medchem Express Supplier No. HY17405

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Alarelin acetate is a synthetic GnRH agonist.

Alarelin acetate is a synthetic GnRH agonist.

Specifications

Chemical Name or Material Alarelin Acetate
CAS 79561-22-1
Color White
Purity Grade Notes Research
Molecular Formula C60H86N16O16
Quantity 10 mg
Synonym Alarelin
Solubility Information DMSO : 100 mg/mL (77.67 mM; Need ultrasonic) ∣H2O : 100 mg/mL (77.67 mM; Need ultrasonic)
SMILES O=C(N[C@@H](CO)C(N[C@@H](CC1=CC=C(O)C=C1)C(N[C@H](C)C(N[C@@H](CC(C)C)C(N[C@@H](CCCNC(N)=N)C(N2CCC[C@H]2C(NCC)=O)=O)=O)=O)=O)=O)[C@@H](NC([C@@H](NC([C@@H](N3)CCC3=O)=O)CC4=CN=CN4)=O)CC5=CNC6=CC=CC=C65.CC(O)=O.CC(O)=O
Molecular Weight (g/mol) 1287.42
Formula Weight 1287.42
Percent Purity 98.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.