Learn More
BINDER™ Alarm Output
Product Code. 30238357 Shop All BINDER Products
Click to view available options
For Use With (Equipment):
KB 65/130/260/470/720/1060
KB PRO Series
KBF PRO Series
KBF Series
Unit Size:
Each
Product Code. 30238357
Brand: BINDER™ 80122370
Only null left
Add to basket
Add to basket
Alarm output, zero-voltage, accessible via 3-pin clamping sleeve
  • Alarm output
  • Zero-voltage
  • Accessible via 3-pin clamping sleeve (max. 24V/2.5A)
  • Acoustic signal that can be switched off
Specifications
For Use With (Equipment) KB 65/130/260/470/720/1060
Material Aluminum
Type Analog Output