missing translation for 'onlineSavingsMsg'
Learn More

Albaconazole, TRC

Product Code. 17977834
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
5 mg
Unit Size:
1mg
2.5mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17977834 1 mg 1mg
17968804 2.5 mg 2.5mg
17937184 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17977834 Supplier Toronto Research Chemicals Supplier No. TRCA5114501MG
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 187949-02-6
Molecular Formula C20 H16 Cl F2 N5 O2
Molecular Weight (g/mol) 431.82
IUPAC Name 7-chloro-3-[(2R,3R)-3-(2,4-difluorophenyl)-3-hydroxy-4-(1,2,4-triazol-1-yl)butan-2-yl]quinazolin-4-one
SMILES C[C@@H](N1C=Nc2cc(Cl)ccc2C1=O)[C@](O)(Cn3cncn3)c4ccc(F)cc4F

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C20H16ClF2N5O2/c1-12(28-11-25-18-6-13(21)2-4-15(18)19(28)29)20(30,8-27-10-24-9-26-27)16-5-3-14(22)7-17(16)23/h2-7,9-12,30H,8H2,1H3/t12-,20-/m1/s1
Formula Weight 431.0961
Chemical Name or Material Albaconazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.