missing translation for 'onlineSavingsMsg'
Learn More

Albendazole Sulfone, TRC

Product Code. 17993465
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17993465 5 mg 5mg
17914595 25 mg 25mg
17932335 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17993465 Supplier Toronto Research Chemicals Supplier No. TRCA5116155MG
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75184-71-3
Molecular Formula C12 H15 N3 O4 S
Molecular Weight (g/mol) 297.33
Synonym Carbamic acid, [5-(propylsulfonyl)-1H-benzimidazol-2-yl]-, methyl ester (9CI), Albendazole sulfone, Methyl [5-(propylsulfonyl)-1H-benzimidazol-2-yl]carbamate
IUPAC Name methyl N-(5-propylsulfonyl-1H-benzimidazol-2-yl)carbamate
SMILES CCCS(=O)(=O)c1ccc2[nH]c(NC(=O)OC)nc2c1

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C12H15N3O4S/c1-3-6-20(17,18)8-4-5-9-10(7-8)14-11(13-9)15-12(16)19-2/h4-5,7H,3,6H2,1-2H3,(H2,13,14,15,16)
Formula Weight 297.0783
Chemical Name or Material Albendazole sulfone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.