missing translation for 'onlineSavingsMsg'
Learn More

Albendazole, TRC

Product Code. 30138055
Change view
Click to view available options
Quantity:
100 mg
1 g
10 g
Unit Size:
100mg
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138055 10 g 10g
30136939 1 g 1g
18050030 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138055 Supplier Toronto Research Chemicals Supplier No. TRCA51161010G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54965-21-8
Molecular Formula C12 H15 N3 O2 S
Molecular Weight (g/mol) 265.33
Synonym Carbamic acid, [5-(propylthio)-1H-benzimidazol-2-yl]-, methyl ester (9CI), Albamelin, Alben, Albendazole, Albendol, Albenza, Albenzil, Aldifal, Alminth, Alvet, Alzental, Andazol, Ashialben, Atasol, Baxen, Bruzol, Eskazole, Exiptol, Haojing, Helminsols, Loveral, Lurdex, Methyl 5-propylthio-2-benzimidazolecarbamate, NSC 220008, SKF 62979, Tobend, Valbazen, Vermisen, Vermitan, Vermizole, Vetalben, Zentel
IUPAC Name methyl N-(5-propylsulfanyl-1H-benzimidazol-2-yl)carbamate
SMILES CCCSc1ccc2[nH]c(NC(=O)OC)nc2c1

Specifications

Quantity 10 g
InChI Formula InChI=1S/C12H15N3O2S/c1-3-6-18-8-4-5-9-10(7-8)14-11(13-9)15-12(16)17-2/h4-5,7H,3,6H2,1-2H3,(H2,13,14,15,16)
Formula Weight 265.0885
Percent Purity >95%
Chemical Name or Material Albendazole
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.