Learn More
Description
Lipid-rich bovine serum albumin for cell culture (1). It is chromatographically purified and has an IgG content ≤ 0.1%.
Origin: New Zealand
Specifications
Specifications
| Content And Storage | Store in refrigerator (2–8°C). |
| Product Type | BSA |
| Product Line | AlbuMAX, Gibco |
| Quantity | 25 g |
| Shipping Condition | Room Temperature |
Frequently Asked Questions (FAQs)
Below are the concentrations of free fatty acids obtained from the HPLC lipid profiles of an older lot of AlbuMAX I:
Alpha-linolenic acid: 0.59 mg FFA/g protein
Linoleic acid: 0.65 mg FFA/g protein
Oleic acid: 2.12 mg FFA/g protein
Stearic acid: 2.09 mg FFA/g protein
Palmitic acid: 1.90 mg FFA/g protein
Note: The individual lipid concentrations may vary between lots, but the values above can be used as a good approximation.
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.