missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ AlbuMax I Lipid Rich BSA

Product Code. 11540376
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11540376 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11540376 Supplier Gibco™ Supplier No. 11020062
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Lipid-rich bovine serum albumin for cell culture. It is chromatographically purified and has an IgG content ≤0.1%. Lyophilized powder.

Lipid-rich bovine serum albumin for cell culture. It is chromatographically purified and has an IgG content ≤0.1%. Lyophilized powder.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 100 g
Storage Requirements 2°C to 8°C
Cell Type Mammalian Cells
Product Type BSA
Sterility Non-sterile
Form Solid
Color Beige

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.