missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ AlbuMax II Lipid Rich BSA

Product Code. 10624053 Shop All Gibco Products
Change view
Click to view available options
Quantity:
5 g
1 kg
5 kg
Unit Size:
1kg
5g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10624053 1 kg 1kg
31781125 5 g 5g
10149072 5 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10624053 Supplier Gibco™ Supplier No. 11021045
Save Up to 60% on Microbiology Essentials
Use promo code 28004
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Lipid-rich bovine serum albumin for cell culture. It is chromatographically purified and has an IgG content ≤1.0%. Lyophilized powder.

This is a lipid-rich bovine serum albumin for cell culture. It is chromatographically purified and has an IgG content ≤ 1%.

Origin: New Zealand

TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 kg
Product Type BSA
Content And Storage Store in refrigerator (2°C to 8°C).
Shipping Condition Room Temperature
Product Line AlbuMAX, Gibco

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.