missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ AlbuMAX™ II Lipid-Rich BSA, 25 g

Product Code. 11550376 Shop All Gibco Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11550376 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11550376 Supplier Gibco™ Supplier No. 11021029
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

AlbuMAX™ II Lipid-Rich BSA

Lipid-rich bovine serum albumin for cell culture (1). It is chromatographically purified and has an IgG content ≤ 1%.

Origin: New Zealand

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in refrigerator (2–8°C).
Cell Type Mammalian Cells
Product Type BSA
Product Line AlbuMAX, Gibco
Quantity 25 g
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.