missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Albumin bovine, for biochemistry, Fraction V, pH=5, 96.0-100.0% protein

Product Code. 10011380 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
100 g
500 g
Unit Size:
100g
10g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10011380 10 g 10g
10326640 100 g 100g
10667794 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10011380 Supplier Thermo Scientific Chemicals Supplier No. 134730100
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Albumin, bovine serum (BSA), CAS # 9048-46-8, is a small, stable biomolecule derived from bovine blood with wide applications in biochemistry, immunochemistry, pharmacokinetics, and cell culture studies. | CAS: 9048-46-8

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description

  • Albumin, bovine serum (BSA) is available as a protease-free, lyophilized powder that may be stably stored at 2-8°c
  • It is comprised of 583 amino acid residues, and has a molecular weight of 66.5 kDa

Applications

  • BSA is used as a protein standard in quantification assays, such as the Bradford protein assay
  • It is used to stabilize enzymes in restriction digestions in molecular biology experiments
  • It is a blocking agent in immunoblots thus increasing signal to noise ratios by quenching non-specific immunochemical reactions
  • BSA is a nutritive agent and used as a supplement in cell and microbial culture media
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9048-46-8
MDL Number MFCD00145743

Specifications

Packaging Glass bottle
Quantity 10 g
Solubility Information Solubility aq. soln.: soluble.
Grade Biochemistry
Chemical Name or Material Albumin bovine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.