missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Albumin, Bovine, For IVD, Low Ash and Metals

Product Code. 11490252 Shop All MP Biomedicals Products
Click to view available options
Quantity:
10 g
25 g
50 g
100 g
500 g
1 kg
Unit Size:
100g
10g
1kg
25g
500g
50g
This item is not returnable. View return policy

Product Code. 11490252

Brand: MP Biomedicals™ 0216006925

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Prepared by heat shock fractionation

  • For IVD applications
  • Low visible absorbance
  • Purity: >98%
  • Ash: <2%
  • Metals: <20ppm

For best cell culture performance, see alternate Mfr. No. 180577

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) ELISA, Microbial Culture
Molecular Weight (g/mol) 66.338
Name Bovine Albumin Fraction V
pH Range 6.5 to 7.5
Quantity 25 g
Source Bovine
Storage Requirements 2° to 8°C
Form Powder
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.