missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Albumin bovine / fraction V, for biochemistry, pH 7.0

Product Code. 10257123 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
100 g
500 g
Packaging:
Glass Bottle
Plastic bottle
Unit Size:
100g
10g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10257123 500 g Glass Bottle 500g
10023060 10 g Plastic bottle 10g
10174520 100 g Plastic bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10257123 Supplier Thermo Scientific Chemicals Supplier No. 240405000
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Albumin bovine/fraction V, CAS # 9048-46-8, also known as bovine serum albumin (BSA), is a serum albumin protein derived from cows. | CAS: 9048-46-8

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Albumin bovine/fraction V is a serum albumin protein commonly used in laboratory experiments including, enzyme-linked immunosorbent assay (ELISAs), immunoblots and immune-histochemistry or -fluorescence

Applications

  • Albumin bovine/fraction V displays blocking activity mainly when casein-sensitive antibodies, including phospho-specific antibodies, are used
  • In genetic approaches this compound can be used to increase polymerase chain reactions (PCR) yields from low purity templates
  • This compound is useful for stabilizing enzymes during storage and also for enzymatic reactions where the absence of nucleases is crucial
  • In in vitro cultures, albumin bovine/fraction V has been applied as a nutrient in cell and microbial cultures especially when serum-free media is used
  • It is commonly used as a protein standard to obtain calibration curves for several laboratorial experiments
  • Albumin bovine/fraction V is a small protein commonly used in immunohistochemistry to block nonspecific regions. The binding of this compound to nonspecific binding sites increases the probability of the antibodies binds only to the region of interest.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9048-46-8
MDL Number MFCD00145743

Specifications

Color Beige-Yellow
Assay Percent Range 96.0% to 100.0% (protein [F:6.22 ]) on dry substance; 98% to 100% (Albumin, 7% solution [by cellulose acetate electrophoresis])
Packaging Glass Bottle
Quantity 500 g
Physical Form Powder
Chemical Name or Material Albumine Bovine/Fraction V

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.