missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(±)-Albuterol-d3 (3-hydroxymethyl-d2; alpha-d1), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | SalbutamolD3(3-hydroxymethyl-D2;aD1) |
| CAS | 1219798-60-3 |
| Molecular Formula | C13 2H3 H18 N O3 |
| Quantity | 0.005 g |
| Synonym | 1,3-Benzenedimethan-α1,α1,α3-d3-ol, α3-[[(1,1-dimethylethyl)amino]methyl]-6-hydroxy-, Salbutamol-D3, Albuterol-D3 |
| InChI Formula | InChI=1 S/C13H21NO3/c1-13(2,3)14-7-12(17)9-4-5-11(16)10(6-9)8-15/h4-6,12,14-17 H,7-8H2,1-3H3/i8D2,12 D |
| SMILES | [2 H]C([2 H])(O)c1cc(ccc1O)C([2 H])(O)CNC(C)(C)C |
| IUPAC Name | 4-[2-(tert-butylamino)-1-deuterio-1-hydroxyethyl]-2-[dideuterio(hydroxy)methyl]phenol |
| Molecular Weight (g/mol) | 242.33 |
| Formula Weight | 242.171 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction