missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Alcian Blue/PAS Special Stain Kit
Sfoglia Tutto Epredia Prodotti
Click to view available options
Type:
Alcian Blue
Alcian Blue/PAS Special Stain Kit
Periodic Acid Schiff
Dimensione della confezione:
500mL
Pack of 3
Descrizione
- Can be used alone to stain acid mucins and other carboxylated and sulfated acid mucosubstances blue on tissue sections
- Periodic Acid Schiff (PAS) reaction is used to stain glycogen, basement membranes and neutral mucosubstances
Includes: Alcian Blue pH 2.5 stain solution, periodic acid solution (0.5%) and Schiff reagent
Specifica
Specifica
| Type | Alcian Blue/PAS Special Stain Kit |
Sicurezza e movimentazione
Identificativo prodotto
- Alcian Blue/PAS Kit 87023
- Danger
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 1
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity after repeated exposure Category 2
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H319-Causes serious eye irritation.
- H371-May cause damage to organs.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P362-Take off contaminated clothing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P310-Immediately call a POISON CENTER/doctor/.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- MIXTURE LIST-Contains : periodic acid, 2-methyl-3-isothiazolone,acetic acid, 5-chloro-2-methyl-3-isothiazolone, copper(4+)
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto