missing translation for 'onlineSavingsMsg'
Learn More

Alcohol, Anhydrous, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Catalog Number 15568604
Click to view available options
Quantity:
4 L
Packaging:
Poly Bottle
This item is not returnable. View return policy
64-17-5
CH4O
32.04
MFCD00004595
OKKJLVBELUTLKV-UHFFFAOYSA-N
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
702
CHEBI:16236
methanol
CO
This item is not returnable. View return policy
64-17-5
CH4O
32.04
MFCD00004595
OKKJLVBELUTLKV-UHFFFAOYSA-N
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
702
CHEBI:16236
methanol
CO

High quality chemicals for laboratory and specialized industrial use

    For Histological Use; NOTE: Intended for nonfuel end purposes only
Grade Meets ACS reagent chemical requirements
Quantity 4 L
Flash Point 13.00°C (55.4°F)
Packaging Poly Bottle
Specific Gravity 1L = 0.79kg
Chemical Name or Material Alcohol, Anhydrous
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H371-May cause damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P405-Store locked up.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'