missing translation for 'onlineSavingsMsg'
Learn More

Tork Alcohol Hand Sanitiser Refill for S4 System

Product Code. 30243863 Shop All Tork Products
Change view
Click to view available options
Form:
Gel
Liquid
Unit Size:
Pack of 6
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Form Unit Size
30243863 Gel Pack of 6
30243831 Liquid Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30243863 Supplier Tork Supplier No. 424103
  • ONLINE EXCLUSIVE PRICE
    115.02 €
    106.71 € / Pack of 6
    Save 8.31 €
    7% Off
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Tork Alcohol Hand Sanitiser is an effective hand sanitiser with 80% denatured alcohol.

  • Help to prevent the spread of infections - this hand sanitiser is fully virucidal and conforms with EN1500, EN12791 and EN14476.
  • Our sanitiser is dermatologically tested and leaves no sticky residue.
  • Help to support better hygiene - this sealed refill bottle has a single-use pump to reduce cross-contamination risk.
  • Reduce waste with a refill bottle that collapses in on itself as it empties.
TRUSTED_SUSTAINABILITY

Specifications

Product Type Hand Sanitizer
Color Clear
Form Gel
Active Ingredients Alcohol
Certifications/Compliance EN1500, EN12791, EN14476
Certifications/Compliance (Europe) EN1500, EN12791, EN14476
Disposable Limited-use
Packaging Plastic Bottle
Quantity 6/Pk.
Volume (Metric) 1 L
compliance-icons compliance-icons
Product Identifier
  • Tork Alcohol Gel Hand Sanitizer
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P264-Wash skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.