Learn More
STEMCELL Technologies ALDEFLUOR™ Assay Buffer
Product Code. 17167711
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mL
55 mL
Unit Size:
25mL
55mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17167711 55 mL 55mL
17157711 25 mL 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17167711 Supplier STEMCELL Technologies Supplier No. 01702
Additional assay buffer for use with the ALDEFLUOR™ kit
  • ALDEFLUOR™ Assay Buffer can be used when counterstaining ALDEFLUOR™-reacted cells with immunofluorescently-labeled antibodies
  • The buffer is necessary to maintain ALDEFLUOR™ reaction product inside the cell, allowing detection of the ALDH-bright population
  • While there is sufficient ALDEFLUOR™-buffer contained within the ALDEFLUOR™ kit, additional buffer may be required for certain immunolabeling procedures
Specifications
For Use With (Application) Cancer Research; Neuroscience; Stem Cell Biology
Quantity 55 mL