missing translation for 'onlineSavingsMsg'
Learn More

Aldosterone 21-Acetate, TRC

Product Code. 30310491
Change view
Click to view available options
Quantity:
2 mg
Unit Size:
2mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30310491 2 mg 2mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30310491 Supplier Toronto Research Chemicals Supplier No. TRCA5147102MG
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifica

Chemical Name or Material Aldosterone 21-Acetate
CAS 2827-21-6
Molecular Formula C23H30O6
Quantity 2 mg
Synonym Aldosterone 21-Monoacetate, (11beta)-21-(Acetyloxy)-11,18-epoxy-18-hydroxy-pregn-4-ene-3,20-dione
InChI Formula InChI=1S/C23H30O6/c1-12(24)28-11-18(26)17-6-5-16-15-4-3-13-9-14(25)7-8-22(13,2)20(15)19-10-23(16,17)21(27)29-19/h9,15-17,19-21,27H,3-8,10-11H2,1-2H3/t15-,16-,17+,19-,20+,21?,22-,23+/m0/s1
SMILES CC(=O)OCC(=O)[C@H]1CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3[C@@H]5C[C@]12C(O)O5
IUPAC Name [2-[(1R,2S,5S,6S,14R,15S,16S)-18-hydroxy-14-methyl-11-oxo-17-oxapentacyclo[14.2.1.01,5.06,15.09,14]nonadec-9-en-2-yl]-2-oxoethyl] acetate
Molecular Weight (g/mol) 402.48
Formula Weight 402.2042
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.