missing translation for 'onlineSavingsMsg'
Learn More

Aldosterone, TRC

Product Code. 30004158
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30004158 1 mg 1mg
30003653 5 mg 5mg
30003398 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30004158 Supplier Toronto Research Chemicals Supplier No. TRCA5147001MG
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52-39-1
Molecular Formula C21 H28 O5
Molecular Weight (g/mol) 360.44
Synonym Pregn-4-en-18-al, 11,21-dihydroxy-3,20-dioxo-, (11β)-, Aldosterone (8CI), (+)-Aldosterone, 11β,21-Dihydroxy-3,20-diketo-4-pregnen-18-al, 11β,21-Dihydroxy-3,20-diketopregn-4-ene-18-al, 11β,21-Dihydroxypregn-4-ene-3,18,20-trione, 18-Formyl-11β,21-dihydroxy-4-pregnene-3,20-dione, 18-Oxocorticosterone, Aldocorten, Aldocortene, Aldocortin, Electrocortin, Elektrocortin, NSC 73856, Reichstein X, d-Aldosterone
IUPAC Name (8S,9S,10R,11S,13R,14S,17S)-11-hydroxy-17-(2-hydroxyacetyl)-10-methyl-3-oxo-1,2,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthrene-13-carbaldehyde
SMILES C[C@]12CCC(=O)C=C1CC[C@H]3[C@@H]4CC[C@H](C(=O)CO)[C@]4(C[C@H](O)[C@H]23)C=O

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C21H28O5/c1-20-7-6-13(24)8-12(20)2-3-14-15-4-5-16(18(26)10-22)21(15,11-23)9-17(25)19(14)20/h8,11,14-17,19,22,25H,2-7,9-10H2,1H3/t14-,15-,16+,17-,19+,20-,21+/m0/s1
Formula Weight 360.1937
Chemical Name or Material Aldosterone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.