missing translation for 'onlineSavingsMsg'
Learn More

Alectinib Hydrochloride, MedChemExpress

Product Code. 16491056
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16491056

Brand: Medchem Express HY13011A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Alectinib Hydrochloride (CH5424802 Hydrochloride; RO5424802 Hydrochloride; AF-802 Hydrochloride) is a potent, selective, and orally available ALK inhibitor with an IC50 of 1.9 nM and a Kd value of 2.4 nM (in an ATP-competitive manner), and also inhibits ALK F1174L and ALK R1275Q with IC50s of 1 nM and 3.5 nM, respectively. Alectinib demonstrates effective central nervous system (CNS) penetration.

Alectinib Hydrochloride (CH5424802 Hydrochloride

Specifications

Chemical Name or Material Alectinib Hydrochloride
CAS 1256589-74-8
Color Off-White
Purity Grade Notes Research
Molecular Formula C30H35ClN4O2
Quantity 200 mg
Synonym CH5424802 HydrochlorideRO5424802 HydrochlorideAF-802 Hydrochloride
Solubility Information DMSO : 5 mg/mL (9.63 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES [H]Cl.N#CC1=CC2=C(C3=C(N2)C(C)(C4=CC(N5CCC(CC5)N6CCOCC6)=C(C=C4C3=O)CC)C)C=C1
Molecular Weight (g/mol) 519.08
Formula Weight 519.08
Percent Purity 98.06%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.