missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ Alexa Fluor™ 350 Antibody Labeling Kit

Product Code. 10654823
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10654823 1 Kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10654823 Supplier Molecular Probes™ Supplier No. A20180

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This kit is optimized for labeling 100 μg of antibody per reaction with blue fluorescent Alexa Fluor™ 350.

Molecular Probes™ Alexa Fluor™ Antibody Labeling Kits provide a convenient means to label small amounts of antibodies with Alexa Fluor™ dyes (choice of 10 colors). This kit is optimized for labeling 100μg of antibody per reaction with blue fluorescent Alexa Fluor™ 350. Comparably small amounts of other proteins (>40 kDa) can also be labeled.

The kit contains everything you need to perform five separate labeling reactions as well as to purify the resulting conjugates. Conjugates are ideal for multiple applications, including flow cytometry, fluorescent microscopy, immunohistochemistry, primary detection, ELISAs, immunocytochemistry, indirect FISH, and more.

Important Features of Alexa Fluor™ 350 Antibody Labeling Kit:
• With an excitation and emission maximum of 346/442 nm, Alexa Fluor™ 350 can be efficiently excited using a UV laser line and detected under standard DAPI filters
• Labeled proteins typically ready to use typically in 90 min (∼15 min hands-on time)
• Useful for labeling 100μg of protein
• Optimized for small-scale labeling of any protein >40 kDa
• Purified using convenient spin filters
• Stabilizing proteins must be removed from the sample before labeling
• Includes detailed instructions for determining degree of labeling (DOL)

Better Results and Workflows with Primary labeled antibodies
A primary antibody directly labeled with a fluorophore often produces lower background fluorescence and less nonspecific binding. Further, multiple primary antibodies of the same isotype or derived from the same species can easily be used in the same experiment if they are directly labeled with compatible fluorophores.

Superior Alexa Fluor™ Dyes
Compared to traditional dyes, Alexa Fluor™ dyes are brighter, more photostable, and more pH resistant between pH 4 and 10. And generally when using Alexa Fluor™ dyes, higher degrees of labeling can be achieved without intramolecular quenching. For details see Alexa Fluor™ Dyes Spanning the Visible and Infrared Spectrum—Section 1.3.

Learn More About Protein and Antibody Labeling
We offer a wide selection of Molecular Probes™ antibody and protein labeling kits to fit your starting material and your experimental setup. See Antibody Labeling from A to Z or use our Labeling Chemistry Selection Tool for other choices. To learn more about our various kits read Kits for Labeling Proteins and Nucleic Acids—Section 1.2 in the Molecular Probes™ Handbook.

We'll Make a Custom Antibody Conjugate for You
If you can't find what you're looking for in our stocked list, we'll prepare a custom antibody conjugate for you. Our custom conjugation service is efficient and confidential, and we stand by the quality of our work. We are ISO 13485:2000 certified.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Labeling Kit
Quantity 1 Kit
Content And Storage Alexa Fluor™ 350 Antibody Labeling Kit comes with 5 vials of reactive dye, sodium bicarbonate, purification resin, spin columns and collections tubes.
  • Store at 2°C to 8°C, protected from light. Component A may be stored from -20°C to +8°C.
  • Detection Method Fluorescence
    Excitation/Emission 346/442 nm
    Labeling Method Conjugation-based
    Labeling Scale 100 μg
    Shipping Condition Room Temperature
    Product Line Alexa Fluor
    Label or Dye Alexa Fluor 350
    Color Blue
    Labeling Target Antibodies, Proteins
    Label Type Alexa Fluor
    Show More Show Less
    I want to use an Alexa Fluor Antibody Labeling kit to label my antibody. The antibody has histidine and glycine in the stock solution. Will this be a problem?

    Yes. The reactive dye in the kit reacts with primary amine groups on histidine and glycine. This can result in less labeling of the antibody. The histidine and glycine must be removed, such as with dialysis or a desalting column. Other components that can interfere include BSA, gelatin, and Tris buffers.

    What formulation of antibody should I use for conjugation for small animal in vivo imaging?

    To allow for good reaction kinetics, antibodies should be in PBS buffer at a concentration of 0.5-3.0 mg/ml. The antibody must be free of preservatives (azide etc.), amine containing buffers and carrier proteins such as BSA.

    What is degree of labeling (DOL)?

    Degree of labeling (DOL) describes the number of fluorophores per antibody. For in vivo labeling experiments, the DOL is restricted to a narrow range because it has significant consequences for the biodistribution and clearance of the probe. For example, for in vivo imaging, we have determined that the DOL range for the far-red Alexa Fluor dyes is 1.5 to 3 molecules per antibody for optimal optical in vivo imaging.

    For Research Use Only. Not for use in diagnostic procedures.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.