missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ Alexa Fluor™ 488 Alkyne (Alexa Fluor™ 488 5-Carboxamido-(Propargyl), Bis(Triethylammonium Salt)), 5-isomer

Product Code. 10287944
Change view
Click to view available options
Quantity:
0.5 mg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10287944 0.5 mg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10287944 Supplier Molecular Probes™ Supplier No. A10267
Powering Proteomics
Use promo code 28003
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Alexa Fluor™ 488 Alkyne (Alexa Fluor™ 488 5-Carboxamido-(Propargyl), Bis(Triethylammonium Salt)), 5-isomer

The green-fluorescent Alexa Fluor™ 488 alkyne is reactive with azides via a copper-catalyzed click reaction. In addition to being an exceedingly bright and photostable fluorophore for use with flow cytometry, microscopy and HCS, Alexa Fluor™ 488 can also be utilized as a bio-orthogonal or biologically unique hapten for use in applications requiring signal amplification.
TRUSTED_SUSTAINABILITY

Specifications

Format Solid
Quantity 0.5 mg
Product Type Alkyne
Molecular Weight (g/mol) 773.91 Da
Solubility DMSO (Dimethylsulfoxide)
Content And Storage Store at ≤-20°C, desiccate and protect from light.
Chemical Reactivity Azide
Detection Method Fluorescence
Reactive Group Alkyne
Shipping Condition Room Temperature
Reactive Moiety Alkyne
Label or Dye Alexa Fluor™ 488
Label Type Alexa Fluor
Color Green
Product Line Alexa Fluor, Molecular Probes
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.