missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ Alexa Fluor™ 555 alkyne, Triethylammonium Salt

Product Code. 10030258
Change view
Click to view available options
Quantity:
0.5 mg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10030258 0.5 mg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10030258 Supplier Molecular Probes™ Supplier No. A20013
 Restricted Item
Estimated Shipment: 19-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Alexa Fluor™ 555 alkyne, triethylammonium salt

The orange-fluorescent Alexa Fluor™ 555 alkyne is designed to react with azide-containing molecules via the fast, selective and extremely efficient copper-catalyzed click reaction.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 0.5 mg
Product Type Alkyne
Content And Storage Store at -20°C, dessicate and protect from light.
Chemical Reactivity Azide
Detection Method Fluorescence
Shipping Condition Room Temperature
System Type Click-iT™
Reactive Moiety Alkyne
Label or Dye Alexa Fluor™ 555
Label Type Alexa Fluor
Color Orange
Product Line Alexa Fluor
Emission 565
Excitation 555
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.