missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Alexa Fluor™ 647-Poly-GT Peptide Substrate

Product Code. 10054044
Click to view available options
Quantity:
1 nmol
10 nmol
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10054044

Brand: Invitrogen™ PV5836

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A general AlexaFluor647-labeled peptide substrate consisting of a polymer of glutamate, and tyrosine that is used to monitor tyrosine kinase acitivty.

A general AlexaFluor647-labeled peptide substrate consisting of a polymer of glutamate, and tyrosine that is used to monitor tyrosine kinase acitivty.

For use in combination with the LanthaScreen™ Eu-PY20 Antibody (Cat. No. PV5696) to evaluate inhibitors in medium to high throughput screening applications.

TRUSTED_SUSTAINABILITY

Specifications

Chromagen Quantity 10nmol
Color Far-Red
Content And Storage AlexaFluor™ 647 Poly-GT peptide substrate in HEPES buffered saline. Store at -20°C.
Detection Method Fluorescence
Form Liquid
For Use With (Application) Pharma and biopharma, target and lead identification and validation, combination with the LanthaScreen™ Eu-PY20 Antibody (PV5696) to evaluate inhibitors in medium to high throughput screening applications, drug discovery and development, kinase biology, biochemical kinase assays, LanthaScreen™ kinase activity assays
Product Line Alexa Fluor™
Quantity 10 nmol
Substrate Random (Glu:Tyr) polymer in a ratio of 4:1
Substrate Properties Peptide-Based Substrate
Substrate Type Kinase Substrate
Target Enzyme Tyrosine Kinases
Excitation/Emission 340/665 nm
Item Description Alexa Fluor™ 647-Poly-GT Peptide Substrate, 10nmol, 30μM Concentration, General Alexa Fluor647-labeled Peptide Substrate Consisting of a Polymer of Glutamate, and Tyrosine that is used to Monitor Tyrosine Kinase Activity
Label or Dye Alexa Fluor™ 647
Product Type Alexa Fluor™ 647-Poly-GT Peptide Substrate
Shipping Condition Dry Ice
Storage Requirements –20°C
Technique TR-FRET
Validated Kinases EGFR
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.