missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Alexa Fluor™ 647-Poly-GT Peptide Substrate

Product Code. 10489864
Change view
Click to view available options
Quantity:
1 nmol
10 nmol
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10489864 1 nmol Each
10054044 10 nmol Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10489864 Supplier Invitrogen™ Supplier No. PV5693
 Restricted Item
Estimated Shipment: 10-08-2026
Añadir a la cesta
Añadir a la cesta
Este artículo no se puede devolver. Vea la política de devoluciones

A general AlexaFluor647-labeled peptide substrate consisting of a polymer of glutamate, and tyrosine that is used to monitor tyrosine kinase acitivty.

A general AlexaFluor647-labeled peptide substrate consisting of a polymer of glutamate, and tyrosine that is used to monitor tyrosine kinase acitivty.

For use in combination with the LanthaScreen™ Eu-PY20 Antibody (Cat. No. PV5696) to evaluate inhibitors in medium to high throughput screening applications.

TRUSTED_SUSTAINABILITY

Especificaciones

Assay Entry Biochemical europium phospho-specific activity
Color Far-Red
Content And Storage 30 μM AlexaFluor 647 Poly-GT peptide substrate in 10 mM HEPES, 137 mM NaCl, 2.7 mM KCl, pH 7.5 (HEPES buffered saline).

Store at -20°C and protect from light.
Detection Method Fluorescence
Form Liquid
Product Line LanthaScreen
Quantity 1 nmol
Substrate Random (Glu:Tyr) polymer in a ratio of 4:1
Substrate Properties Peptide-Based Substrate
Substrate Type Kinase Substrate
Target Enzyme Tyrosine Kinases
Excitation/Emission 340/665 nm
Label or Dye Alexa Fluor™ 647
Shipping Condition Dry Ice
Target Entry ABL1 G250E, CSF1R, CSK, EPHA5, KIT, PDGFRA V561D, SYK, TYRO3, ZAP70, FGFR1, FGFR2, EGFR, EPHB2, FGFR4, IGF1R, LYNA, MATK, MERTK, NTRK3, RET V804L, TEK, FLT1, MUSK, SRCN1, ABL1 E255K, EPHA4, FYN, PTK2, PTK2B, RET, ABL1 T315I, EPHA3, EPHB4, FLT3, FLT4, INSR, LCK, YES1, EGFR L858R, ERBB4, KDR, MET, TNK2, ABL1, ABL2, AXL, FGFR3, FGR, HCK, KIT V654A, EPHA1, EPHA8, EPHB3, INSRR, LYNB, NTRK1, PDGFRA D842V, SRMS, EGFR L861Q, EPHA2, FER, LTK, PDGFRA, EPHA7, EPHB1, FRK, ITK, FLT3 D835Y, SRC, BTK, FES, ERBB2, NTRK2
Technique TR-FRET
Validated Kinases EGFR
Mostrar más Mostrar menos

For Research Use Only. Not for use in diagnostic procedures.

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.