missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Alexa Fluor™ 700 NHS Ester (Succinimidyl Ester)

Product Code. 10072682
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10072682 1 mg 1mg
11554017 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10072682 Supplier Invitrogen™ Supplier No. A20010

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Alexa Fluor™ 700 is a bright and photostable near-IR dye that can be excited with a xenon-arc lamp, far-red diode lasers, or dye pumped lasers operating in the 675–700 nm range.

Alexa Fluor™ 700 is a bright and photostable near-IR dye that can be excited with a xenon-arc lamp, far-red diode lasers, or dye pumped lasers operating in the 675–700 nm range. Used for stable signal generation in imaging and flow cytometry, Alexa Fluor™ 700 dye is water soluble and pH-insensitive from pH 4 to pH 10. Fluorescence of this long-wavelength Alexa Fluor™ dye is not visible to the human eye but is readily detected by most imaging systems. In addition to reactive dye formulations, we offer Alexa Fluor™ 700 dye conjugated to a variety of antibodies, peptides, proteins, tracers, and amplification substrates optimized for cellular labeling and detection.The NHS ester (or succinimidyl ester) of Alexa Fluor™ 700 is the most popular tool for conjugating this dye to a protein or antibody. NHS esters can be used to label to the primary amines (R-NH2) of proteins, amine-modified oligonucleotides, and other amine-containing molecules. The resulting Alexa Fluor™ conjugate will exhibit brighter fluorescence and greater photostability than the conjugates of other spectrally similar fluorophores.

Detailed information about this AlexaFluor™ NHS ester:
Fluorophore label: Alexa Fluor™ 700 dye
Reactive group: NHS ester
Reactivity: Primary amines on proteins and ligands, amine-modified oligonucleotides
Ex/Em of the conjugate: 702/723 nm
Extinction coefficient: 205,000 cm-1M-1
Molecular weight: ∼1400

Typical Conjugation Reaction
You can conjugate amine-reactive reagents with virtually any protein or peptide (the provided protocol is optimized for IgG antibodies). You can scale the reaction for any amount of protein, but the concentration of the protein should be at least 2 mg/mL for optimal results. We recommend trying three different degrees of labeling, using three different molar ratios of the reactive reagent to protein.

The Alexa Fluor™ NHS ester is typically dissolved in high-quality anhydrous dimethylformamide (DMF) or dimethylsulfoxide (DMSO) (D12345), and the reaction is carried out in 0.1–0.2 M sodium bicarbonate buffer, pH 8.3, at room temperature for 1 hour. Because the pKa of the terminal amine is lower than that of the lysine epsilon-amino group, you may achieve more selective labeling of the amine terminus using a buffer closer to neutral pH.

Conjugate Purification
Labeled antibodies are typically separated from free Alexa Fluor™ dye using a gel filtration column, such as Sephadex™ G-25, BioGel™ P-30, or equivalent. For much larger or smaller proteins, select a gel filtration media with an appropriate molecular weight cut-off or purify by dialysis. We offer several purification kits optimized for different quantities of antibody conjugate:
Antibody Conjugate Purification Kit for 0.5-1 mg (Cat. No. A33086)
Antibody Conjugate Purification Kit for 20-50 μg (Cat. No. A33087)
Antibody Conjugate Purification kit for 50-100 μg (Cat. No. A33088)

Learn More About Protein and Antibody Labeling
We offer a wide selection of Molecular Probes™ antibody and protein labeling kits to fit your starting material and your experimental setup. See our Antibody Labeling kits or use our Labeling Chemistry Selection Tool for other choices. To learn more about our labeling kits, read Kits for Labeling Proteins and Nucleic Acids—Section 1.2 in The Molecular Probes™ Handbook.

We’ll Make a Custom Conjugate for You
If you can’t find what you’re looking for in our online catalog, we’ll prepare a custom antibody or protein conjugate for you. Our custom conjugation service is efficient and confidential, and we stand by the quality of our work. We are ISO 13485:2000 certified.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 mg
Product Type Dye
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Chemical Reactivity Amine
Shipping Condition Room Temperature
Reactive Moiety Active Ester, Succinimidyl Ester
Label or Dye Alexa Fluor™ 700
Label Type Alexa Fluor
Product Line Alexa Fluor
Emission 723 nm
Excitation 702 nm
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.