missing translation for 'onlineSavingsMsg'
Learn More

Alexidine-d10 Dihydrochloride, TRC

Product Code. 30314030
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30314030 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30314030 Supplier Toronto Research Chemicals Supplier No. TRCA52500210MG
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Alexidine-d10 Dihydrochloride
CAS 1246818-47-2
Molecular Formula C26H48D10Cl2N10
Quantity 10 mg
Synonym N,N''-Bis[2-(ethyl-d5)hexyl]-3,12-diimino-2,4,11,13-tetraazatetradecanediimidamide, 1,1'-Hexamethylene-bis[5-[2-(ethyl-d5)hexyl]biguanide], Bisguadine-d10, Bisguanidine-d10, Compound 904-d10, QR 711-d10, Sterwin 904-d10, Win 21904-d10
InChI Formula InChI=1S/C26H56N10/c1-5-9-15-21(7-3)19-33-25(29)35-23(27)31-17-13-11-12-14-18-32-24(28)36-26(30)34-20-22(8-4)16-10-6-2/h21-22H,5-20H2,1-4H3,(H5,27,29,31,33,35)(H5,28,30,32,34,36)/i3D3,4D3,7D2,8D2
SMILES N=C(NC(NCC(C([2H])([2H])C([2H])([2H])[2H])CCCC)=N)NCCCCCCNC(NC(NCC(C([2H])([2H])C([2H])([2H])[2H])CCCC)=N)=N.Cl.Cl
IUPAC Name 1-[N'-[6-[[amino-[[N'-[2-(1,1,2,2,2-pentadeuterioethyl)hexyl]carbamimidoyl]amino]methylidene]amino]hexyl]carbamimidoyl]-2-[2-(1,1,2,2,2-pentadeuterioethyl)hexyl]guanidine
Molecular Weight (g/mol) 591.77
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.