Learn More
Thermo Scientific Chemicals Zinc, Quantofix™ Test Strips
Brand: Thermo Scientific Chemicals 046651.QA
83.33 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.01000kg Transport : UN number : 1824 Chem class : 8 Pack group : III
Alert:
H314; Causes severe skin burns and eye damage. P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a; Do not breathe dust/fume/gas/mist/vapours/spray. IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF SWALLOWED: Rinse mouth. Do NOT induce vomiting. Store locked up. Dispose of contents/container in accordance with local/regional/national/international regulations.
Description
Zinc, Quantofix™ Test Strips is used to measure the determination of zinc in solutions quickly. It can be used readily for measurements. It is used to regularly check the plating baths in the metal plating industry. It is also used to check the concentration of zinc in cooling water treatment, where zinc salts are used as corrosion inhibitors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Liquid
- Colorless
- Fully miscible
Specifications
Liquid | |
Reagents and 100 strips to measure 0-100ppm Zn+2 in aqueous solution | |
Fully Miscible |
RUO – Research Use Only