missing translation for 'onlineSavingsMsg'
Learn More

Alfuzosin, MedChemExpress

Product Code. 16488398
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16488398 10 mg 10mg
16474287 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16488398 Supplier Medchem Express Supplier No. HYB0192
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Alfuzosin is an α1 adrenergic receptor antagonist used to treat benign prostatic hyperplasia (BPH).

Alfuzosin is an α1 adrenergic receptor antagonist used to treat benign prostatic hyperplasia (BPH).

Specifications

Chemical Name or Material Alfuzosin
CAS 81403-80-7
Color White
Purity Grade Notes Research
Molecular Formula C19H27N5O4
Quantity 10 mg
Synonym SL 77499
Solubility Information DMSO : ≥ 50 mg/mL (128.39 mM)
SMILES O=C(C1OCCC1)NCCCN(C2=NC(N)=C3C=C(OC)C(OC)=CC3=N2)C
Molecular Weight (g/mol) 389.45
Formula Weight 389.45
Percent Purity 99.67%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.