missing translation for 'onlineSavingsMsg'
Learn More

Alfuzosin, TRC

Product Code. 30008041
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
100mg
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30008041 100 mg 100mg
30004276 500 mg 500mg
30011773 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30008041 Supplier Toronto Research Chemicals Supplier No. TRCA215435100MG
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 81403-80-7
Molecular Formula C19H28ClN5O4
Molecular Weight (g/mol) 425.91
Synonym 2-Furancarboxamide, N-[3-[(4-amino-6,7-dimethoxy-2-quinazolinyl)methylamino]propyl]tetrahydro-, N-[3-[(4-Amino-6,7-dimethoxy-2-quinazolinyl)methylamino]propyl]tetrahydro-2-furancarboxamide, Alfuzosin, N-[3-[N-(4-Amino-6,7-dimethoxyquinazolin-2-yl)-N-(methyl)amino]propyl]tetrahydrofuran-2-carboxamide, SL 77499
IUPAC Name N-[3-[(4-amino-6,7-dimethoxyquinazolin-2-yl)-methylamino]propyl]oxolane-2-carboxamide
SMILES COc1cc2nc(nc(N)c2cc1OC)N(C)CCCNC(=O)C3CCCO3

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C19H27N5O4/c1-24(8-5-7-21-18(25)14-6-4-9-28-14)19-22-13-11-16(27-3)15(26-2)10-12(13)17(20)23-19/h10-11,14H,4-9H2,1-3H3,(H,21,25)(H2,20,22,23)
Formula Weight 389.2063
Chemical Name or Material Alfuzosin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.