missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Alginic acid, sodium salt

Product Code. 10062400 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
250 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic Bottle
Unit Size:
2.5kg
250g
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10062400 250 g Plastic Bottle 250g
10521631 5 g Glass bottle 5g
10215710 500 g Plastic Bottle 500g
11936991 2.5 kg Plastic Bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10062400 Supplier Thermo Scientific Chemicals Supplier No. 177772500
 Restricted Item
Estimated Shipment: 15-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 9005-38-3 | (C6H7O7)A(C6H7O7)BNa

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9005-38-3
Molecular Formula (C6H7O7)A(C6H7O7)BNa
MDL Number MFCD00081310
Synonym Algin, Sodium alginate

Specifications

Quantity 250 g
Packaging Plastic Bottle
Chemical Name or Material Alginic acid, sodium salt
What is the molecular weight and M/G ratio of Alginic acid, sodium salt (Cat. No. 177775000, 177770050, 177772500, 177770025)?

The molecular weight of Alginic acid, sodium salt (Cat. No. 177775000, 177770050, 177772500, 177770025) is 450,000-550,000 Dalton and the M/G ratio is 1.5-1.6, thus ~60% mannuronic acid (m) and ~40% guluronic acid (n).

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.